CAS RN 1224846-01-8
CAS RN 1224846-01-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MK-3697, MedChemExpress
MedChemExpress MK-3697 is an isonicotinamide small molecule, acting as a potent and selective Orexin 2 receptor antagonist with Ki = 0.95 nM.