CAS RN 120511-92-4
CAS RN 120511-92-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2'-[5-(4H-1,2,4-Triazol-4-ylmethyl)benzene-1,3-diyl]bis(2-methylpropanenitrile) (Isoanastrozole), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.