CAS RN 117428-22-5
CAS RN 117428-22-5
IUPAC Name:
methyl 3-methoxy-2-[2-({[6-(trifluoromethyl)pyridin-2-yl]oxy}methyl)phenyl]prop-2-enoate
Synonyms:
methyl 3-methoxy-2-[2-({[6-(trifluoromethyl)pyridin-2-yl]oxy}methyl)phenyl]prop-2-enoate
Molecular Weight (g/mol):
367.32
Molecular Formula:
C18H16F3NO4
InChi Key:
IBSNKSODLGJUMQ-UHFFFAOYSA-N
SMILES:
COC=C(C(=O)OC)C1=CC=CC=C1COC1=CC=CC(=N1)C(F)(F)F
1
–
1
of
1
results
1
Picoxystrobin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality