CAS RN 1173998-58-7
CAS RN 1173998-58-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(2alpha,3alpha,5alpha,17beta)-2,17-Dimethylandrostane-3,17-diol, TRC
CAS: 1173998-58-7 Molecular Formula: C21 H36 O2 Molecular Weight (g/mol): 320.51 Synonym: (2α,3α,5α,17β)-2,17-Dimethylandrostane-3,17-diol,2α,17α-Dimethyl-5α-androstane-3α,17β-diol IUPAC Name: (2R,3S,5S,8R,9S,10S,13S,14S,17S)-2,10,13,17-tetramethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthrene-3,17-diol SMILES: C[C@@H]1C[C@@]2(C)[C@@H](CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(C)O)C[C@@H]1O
2
2alpha,17alpha-Dimethyl-5alpha-androstane-3alpha,17beta-diol, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.