CAS RN 1150310-15-8
CAS RN 1150310-15-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2E)-3-(3,4-Dihydroxy-5-nitrophenyl)-2-(piperidin-1-ylcarbonyl)prop-2-ennitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.