accessibility menu, dialog, popup

UserName

CAS RN 109903-35-7

CH3HNSOONH2

CAS RN 109903-35-7

IUPAC Name: 1-(4-aminophenyl)-N-methylmethanesulfonamide
Synonyms: 1-(4-aminophenyl)-N-methylmethanesulfonamide
Molecular Weight (g/mol): 200.26
Molecular Formula: C8H12N2O2S
InChi Key: CIWNHTXCBHTWRV-UHFFFAOYSA-N
SMILES: CNS(=O)(=O)CC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results