accessibility menu, dialog, popup

UserName

CAS RN 1074-43-7

CH3H3C

CAS RN 1074-43-7

IUPAC Name: 1-methyl-3-propylbenzene
Synonyms: 1-methyl-3-propylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: QUEBYVKXYIKVSO-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC(C)=C1
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results