Fisher Scientific UK Ltd.
Bishop Meadow Road,
Loughborough,
LE11 5RG
Certificate of Analysis
Thermo Fisher Scientific's Quality System has been found to conform to Quality Management System Standard ISO9001:2015 by SAI Global Certificate Number. QMS42099
Catalogue Number 40682
Lot Number 2108234
Description 1,2-Dichloroethane , ACS meets req.of Reag.Ph.Eur.
Quality Test / Release Date 23-DEC-21
Expiry Phrase Use within 3 years of opening
Country of Origin Germany
Declaration of Origin Synthetic
BSE/TSE Not directly derived from or manufactured with any animal byproducts in any way (Including but not limited to fermentation or nutrient broth, catalysts, enzymes).
Result Name Units Specification Test Value
Appearance Clear colourless Mobile liquid Clear colourless Mobile liquid
Assay (GC) % >= 99.0
99.99
Colour APHA <= 10
<10
Density (g/ml) g <= 1.25
1.253
Residue after evaporation (ppm) ppm <= 20
3.2
Titrable acid meq/g <= 0.0003
0.00008
Water % <= 0.03
0.004
Certified By
Mrs Paula Faulkes Wednesday 17th of December 2025
Quality Assurance Manager.
Products are processed under ISO 9001:2015 quality management systems and samples are tested for conformance to the noted specifications. Certain data may have been supplied by third parties. We disclaim the implied warranties of merchantability and fitness for a particular purpose, and the accuracy of third-party data or information associated with the product. Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator or end user to determine suitability, and to qualify and/or validate each product for its intended use.