missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Solid Top Screw Caps
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
£78.10 - £221.00
Specifications
Product Type | Screw Cap |
---|---|
Liner Material | Styrene Butadiene Rubber |
Autoclavable | Autoclavable |
Material | Phenolic |
Color | Black |
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
12415081
|
DWK Life Sciences
W239298 |
20-400 |
£78.20
Pack of 144 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10423332
|
DWK Life Sciences
240215 |
18-400 |
£221.00
Pack of 500 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
12435951
|
DWK Life Sciences
W239300 |
24-400 |
£97.55
Pack of 144 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
10130721
|
DWK Life Sciences
240209 |
15-425 |
£78.10
Pack of 200 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
Description
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
- 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
- Aluminum foil faced pulp board liners can be used with organic solvents
- Use LDPE lined caps with distilled water, analytical standards and reagents
Specifications
Screw Cap | |
Autoclavable | |
Black |
Styrene Butadiene Rubber | |
Phenolic | |
Ideal for cell culture work |