missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ UniPrep™ Syringeless Filters - Nylon

Process samples in one-third the time of conventional syringe filters

Brand:  Cytiva UN113UNYL

Code : 69

Additional Details : Weight : 0.43000kg

 View more versions of this product

Product Code. 11348724

  • £231.00 / Pack of 50
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

UniPrep syringeless filters are preassembled filtration devices for the filtration and storage of laboratory samples.

  • Naturally hydrophilic membrane for filtration of samples containing aqueous or organic solvents with a pH range between 3 to 10
  • Consists of external sample test tube, plunger (which incorporates membrane filter and prefilter), removable cap
  • Filtration area: 0.3cm2
  • Safe, efficient—prefilter and membrane can't pop off
  • Air purge unnecessary—inserting plunger into tube provides a positive pressure purge of sample through filter
  • Sealed glass microfiber prefilter traps larger particles before they can clog the membrane

  • Increase lab productivity and efficiency
  • Generate less waste
  • Realize greater overall cost savings per test

Specifications

Specifications

Nylon
0.28 cm2
Pk. of 50
17 mm
0.45 μm
50 μL
Syringeless Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers