missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Vacuum Filtration Holder
Optimize your vacuum filtration process with the Vacuum Filtration Holder.
£265.00 - £269.00
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Equipment) | Vacuum Filter Holder |
Material | Aluminum |
Quantity | 1 Ea. |
Type | Vacuum Filtration Holder |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11721883
|
Cole-Parmer™
02921-00 |
15 mL |
£265.00
Each |
In Stock
Log in to see stock. |
|||||
11711883
|
Cole-Parmer™
02920-00 |
100 mL |
£269.00
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
- Model 02923-20 meets EPA specifications for Hazardous Waste Toxicity Test.
- Autoclavable.
- Supplied with an aluminum clamp and a silicone rubber stopper.
- No. 5 stoppers fit standard 125 mL flasks.
- No. 8 stoppers fit 1 L flasks.
- Order no. 8B stopper 02924-02 separately to fit filter holders 02920-00 and 02921-00 to manifolds above.
- Filter holder 02923-10 is ideal for sterility testing of antibiotics and pharmaceuticals by bacteria retentive method described in USP XXI.
- This model includes funnel, filter base, aluminum clamp, and stopper.
- Model 02923-20 with sintered-glass membrane support meets EPA specifications for use in the Hazardous Waste Toxicity Test.
- Holder 02923-00, with stainless steel (SS) membrane support, provides ultraclean filtrate—screen does not shed particles.
Specifications
Autoclavable | |
Aluminum | |
Vacuum Filtration Holder |
Vacuum Filter Holder | |
1 Ea. |