Learn More
Jaytec Glass™ Type 3 Gay-Lussac Style Borosilicate Glass Density Bottles
Type 3 Gay-Lussac Style Borosilicate Glass Density Bottles are used to determine the density of liquids of moderate viscosity.
Brand: Jaytec Glass™ WJ765
Code : 02
Additional Details : Weight : 0.01000kg
Description
- Borosilicate glass
Specifications
5 x 25 mL | |
To determine the density of a liquid | |
Density Bottle | |
Borosilicate Glass |