missing translation for 'onlineSavingsMsg'
Learn More

Tripotassium citrate monohydrate, 99+%

Product Code. p-7050005
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337538 500 g 500g
11327538 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337538 Supplier Thermo Scientific Alfa Aesar Supplier No. 033222.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6100-05-6 | C6H7K3O8 | 324.41 g/mol

Tripotassium citrate hydrate is used in food and beverage industries. It finds application as buffering, sequestering or emulsifying agent. It is also used as a potassium source in pharmaceuticals and an active ingredient to treat urinary duct stones. It is added to oral care products to reduce the teeth sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
MDL Number MFCD00150442
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Melting Point 230°C (decomposition)
Density 1.98
Assay Percent Range >99%
Quantity 500 g
Merck Index 14,7623
Solubility Information Soluble in water and glycerol; Practically insoluble in alcoholSoluble in water and glycerol; Insoluble in alcohol
Formula Weight 324.42 (306.41 Anhydrous)
Percent Purity ≥99%
Physical Form Granules
Chemical Name or Material Tripotassium citrate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.