missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid, BAKER ANALYZED™ Reagent, J.T. Baker™
755.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Formula Weight | 114.03 |
Grade | BAKER ANALYZED™ Reagent |
Quantity | 500 mL |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.53kg |
Chemical Name or Material | Trifluoroacetic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'