missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine, 99+%, Thermo Scientific Chemicals
13.52 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Use promo code "21742" to get your promotional price.
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 1.1245g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Glass Bottle |
Molecular Formula | C6H15NO3 |
Linear Formula | (HOCH2CH2)3N |
Refractive Index | 1.4830 to 1.4860 (20°C, 589nm) |
Quantity | 100 mL |
Fieser | 01,1196 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'