missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Tricorn™ Mono™ Q HR 16/10 Prepacked IEX Columns
Strong anion exchange media, pre-packed in columns, designed for high performance, laboratory scale separations of proteins and other biomolecules based on charge or isoelectric point
Brand: Cytiva 17-0506-01
Code : GX
Additional Details : Weight : 0.35600kg
Description
- Fast, efficient, and cost-effective polishing step in purification of proteins, peptides etc. when purity is required
- Ready-to-use columns for convenient process development optimization
- Monodisperse porous beads for high resolution purification and high capacity
- The 20 ml bed volume of the column offers high capacity
- Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
Specifications
All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid | |
20mL | |
16mm | |
3.93 in. | |
10mL/min. | |
O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3 | |
2 to 12 | |
Monodisperse porous Polystyrene/Divinyl Benzene bead | |
Ion Exchange Column |
Mono Q HR monobead prepacked IEX column | |
0.62 in. | |
1 x 20mL | |
100mm | |
Borosilicate Glass | |
435psi | |
10μm | |
4° to 30°C | |
Polishing step in purification of Proteins, Peptides and other biomolecules when high purity is required |