Learn More
Invitrogen™ eBioscience™ TMB Solution (1X)
Recommended for ELISA Ready-SET-Go!™ kits. eBioscience™ TMB ELISA Substrate Solution, 1x is a sensitive chromogenic substrate supplied as single ready-to-use solution.
Brand: Invitrogen™ 00-4201-56
Code : NEW
Additional Details : Weight : 0.01000kg
Description
TMB (3,3’,5,5’-Tetramethylbenzidine) is a substrate for horseradish peroxidase (HRP). During the enzymatic reaction the TMB substrate will result in a blue color that can be read on a spectrophotometer at 650nm. This kinetic reaction can be stopped using 1M phosphoric acid resulting in a color change to yellow that can be read at 450nm.
Specifications
TMB Solution | |
eBioscience™ | |
HRP (Horseradish Peroxidase) |
Colorimetric | |
Chemical Substrate |
For Research Use Only. Not for use in diagnostic procedures.