missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thionyl chloride, 99.7%, Thermo Scientific Chemicals
43.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -105.0°C |
Density | 1.6400 g/mL |
Boiling Point | 76.0°C |
Assay Percent Range | 99.7% |
Packaging | Glass bottle |
Linear Formula | SOCl2 |
Refractive Index | 1.5190 to 1.5210 |
Quantity | 1 L |
Merck Index | 15, 9501 |
Specific Gravity | 1.64 |
Show More |
Safety and Handling
Product Identifier
- Thionyl chloride
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'