missing translation for 'onlineSavingsMsg'
Learn More

JULABO TE400 Peltier chiller

With their robust design and precise temperature control, Peltier chillers are ideal for simple temperature control tasks in laboratories and research facilities.

Brand:  JULABO 9635400

Additional Details : Weight : 23.75000kg

Product Code. 17927821

  • £6081.75 / Each
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Operate without refrigerants and are therefore very environmentally friendly. At the same time, the reliable chillers contribute to lowering operating costs and reducing the ecological footprint thanks to their energy-efficient operation.
  • The TE400 uses fewer movable components such as compressors for cooling. This makes it comparatively low-maintenance and less susceptible to wear. The Peltier chiller offers a cooling capacity of up to 400W and a pump capacity of up to 7 L/min and impresses with its high temperature stability for a wide range of different applications.
  • High temperature stability
  • Environmentally friendly operation with low energy consumption
  • Economical operation due to high energy efficiency and low energy consumption
  • Touch display for easy operation
Specifications

Specifications

1 L
Air-cooled
7 L/min.
Centrifugal Pump
Recirculating Chiller
RS232
0.4kW at 20 °C
Ideal for simple temperature control tasks in laboratories and research facilities.
-5°C to +40°C
100/230 V 50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers