Learn More
Agilent BioTek Synergy H1 Hybrid Multi-Mode Reader
BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. For Research Use Only. Not for use in diagnostic procedures.
Brand: Agilent BioTek SH1M-SN
Description
BioTek Synergy H1 is a modular multi-mode microplate reader, with monochromator-based optics and filter-based optics. The patented Hybrid Technology offers flexibility and sensitivity across a broad range of applications. The modular platform allows upgrading to expand functionality as your laboratory workflows change
Synergy H1 offers continuously variable bandwidth monochromators for fluorescence excitation and emission wavelength selection. Fluorescence bandwidth can be set between 9 and 50 nm, in 1 nm increments, allowing users to fully optimize reader settings to drive the best assay performance compared to fixed bandwidth systems.
Specifications
230 to 999 nm (1 nm increments) | |
CE and TUV marked. RoHS compliant. | |
46.4 cm | |
18.3 in. | |
11 sec. (96 wells), 22 sec. (384 wells) | |
0 to 4 OD | |
13 in. | |
33 cm | |
Proprietary Hybrid Technology | |
2 Syringe Pumps | |
130 W max. | |
2.5 to 4 pM monochromators, 0.25 pM filters fluorescence, 20 amol ATP monochromator, 10 amol ATP filter luminescence, 1.2 mP fluorescence polarization, Europium 40 fM with filters, 1200 fM with monos time resolved fluorescence | |
<55 lb. | |
14.8 in. | |
6- to 384-well Plates, Take3 Micro-volume Plates | |
Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
End Point, Kinetic, Spectral Scanning, Well Area Scanning |
Absorbance: ±2 nm; Dispenser: ±1 μL or 2% | |
0 to 20% | |
BioStack and Third Party Automation Compatible | |
4-Zone Incubation to 45°C | |
Linear, Orbital, Double Orbital | |
5 to 1000 μL, 1 μL increments | |
50/60 Hz | |
Monochromator fluorescence; Monochromator absorbance; Full-light luminescence; Time resolved fluorescence | |
H1M | |
Multi-Mode Microplate Reader | |
±0.1% (CO2 and O2) | |
Fluorescence Intensity Monochromator: 250 to 700 nm (900 nm option), Fluorescence Polarization: 280 to 700 nm (850 nm option) | |
25 kg | |
37.5 cm | |
Photodiode, PMT | |
Xenon Flash |