missing translation for 'onlineSavingsMsg'
Learn More

BUSE Scientific Sulfur Hexafluoride Gas

Brand:  BUSE Scientific LBSULH30

Code : 22

Additional Details : CAS Number : 2551-62-4 Weight : 0.22700kg Transport : UN number : 1080 Chem class : 2.2

Product Code. 12999534

  • £281.43 / 227g
Estimated Shipment: 11-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Sulfur Hexafluoride
227 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions