missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP Bel-Art™ Sterileware Jumbo Sampling Pipettes
Ideal for sampling from narrow- or wide-mouth containers up to 208-liter (55 Gallon) drums
£378.00 - £510.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
16876916
|
SP Bel-Art™
F37872-0450 |
80 mL |
£378.00
Pack of 50 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
16886916
|
SP Bel-Art™
F37872-0900 |
180 mL |
£510.00
Pack of 50 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
16896916
|
SP Bel-Art™
F37872-1100 |
225 mL |
£378.00
Pack of 37 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
- Reaches into container while keeping hand clear of contents
- Diameter allows for use through drum bunghole or other small openings
- Color-coded end helps prevent handling errors
- Low-viscosity liquids enter easily through the 3 mm (1⁄8 in.) tip hole
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) pipettes heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
2.1 cm |