missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Sterile PES Syringe Filter
Designed for filtration and purification in a sterile environment. Fisherbrand™ Sterile PES Syringe Filter features a PES membrane in a polypropylene housing for purification of tissue culture solutions from a standard syringe. 33mm diameter membranes available with 0.2 or 0.45μm pore sizes.
£99.50 - £105.26
Specifications
Color | White |
---|---|
Filter Type | PES |
Sterility | Sterile |
For Use With (Application) | Purification of tissue culture solutions, buffers |
Housing Material | Polypropylene |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
15206869
|
Fisherbrand™
|
0.2 μm |
£99.50
Pack of 50 |
In Stock
Log in to see stock. |
|||||
15216869
|
Fisherbrand™
|
0.45 μm |
£105.26
Pack of 50 |
In Stock
Log in to see stock. |
|||||
Description
- Ideal for purification of tissue culture solutions and buffers
- High sample throughput with low protein binding
- Hydrophilic 33mm polyethersulfone (PES) membrane
- Sturdy, chemically resistant polypropylene housing
Specifications
White | |
Sterile | |
Polypropylene | |
Syringe Filter |
PES | |
Purification of tissue culture solutions, buffers | |
50 Pack | |
33 mm |