missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Steel O-ring

£11.00 - £17.00

Specifications

Product Type O-ring
Material Steel
Packaging Quantity 1 Ea.
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11924352
View Documents Promotion Details
RETSCH
05.114.0075
For Vibratory Disc Mill RS 200 50 mL Grinding Set
£11.00
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
10208841
View Documents Promotion Details
RETSCH
05.114.0076
For Vibratory Disc Mill RS 200 250 mL Grinding Set
£17.00
Each
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

O-ring
Steel
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications