Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
Brand: Fisherbrand™
Additional Details : Weight : 2.00000kg
Description
- Class F2 stainless steel calibration weight
- Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
F2 | |
1 g to 1 kg | |
1 g to 1 kg | |
Cylindrical |