Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F1 with certificate
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
£886.16 - £2581.10
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
15535109
|
Fisherbrand™
|
1 to 100 g |
£886.16
1 unit |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15595129
|
Fisherbrand™
|
1 g to 5 kg |
£2581.10
1 unit |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15515129
|
Fisherbrand™
|
1 g to 1 kg |
£1409.80
1 unit |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
- Class F1 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
F1 |