Learn More
Specac™ Specamill™ Stainless Steel Capsule Set
Brand:  Specac™ GS06100
Code : Z3
Additional Details : Weight : 0.04000kg
Product Code. 15240048
  • £211.75 / Each
Estimated Shipment: 13-06-2024
to see stock.
Description
Description
Stainless Steel Capsule set for use in GS06000 and GS06001. This set is typically of a Mohs hardness of 7.5 and ideal for hard samples. Contains Stainless Steel stopper and 3 stainless steel ball bearings.
Specifications
Specifications
GS06000 and GS06001
Stainless Steel stopper and 3 stainless steel ball bearings