Learn More
Faster™ SmartFAST Laminar Flow Hood
SmartFAST Cabinets are the new concept of compact ISO 3 vertical laminar flow cabinets, which guarantee a decontaminated working area and particle-free conditions. They are especially designed for molecular biology applications.
Brand:  Faster™ F00012100000
Additional Details : Weight : 150.00000kg
Product Code. 16288470
  • £4675.00 / Each
Estimated Shipment: 16-07-2024
to see stock.
Description
Description
Ergonomic Design: The angled sloping (7°) front hinged safety-glass provides optimum visibility of all objects placed in the interior workspace. The frontal opening is set to 200 mm. The whole front of the cabinet can also be opened upwards as it is hinged on the top – in order to allow easy access for complete and effective cleaning and decontamination.
Economic and easy-to-place: SmartFAST are the compact version of vertical laminar flow cabinet with overall width dimension of 830 mm that allow to place the cabinet also in restricted spaces.
ECS™ Eco Controling System: The new ECS™ microprocessor employs the latest innovative methods of integrated management of all principal functions of ventilation and filtration - self-regulating all the main filtration and ventilation system components - compensating for declining pressure drops and restoring power balance. Combining the use of AC motor-blower and certified low pressure-drop filters, the new ECS™ controlling system optimize power consumption, reducing CO2 emissions into the environment.
Anti Bacterial Coating: Exclusive Dupont™ ALESTA™ anti-bacterial 'Ag+ cations-based solution', capable to prevent microbial contamination of surfaces thereby inhibiting long term surface growth. Made of Stainless Steel AISI 304 L.
High Level Lighting: The safety glass side-windows with the ideal positioning and sizing of the light-system provide the highest level of luminosity to the work area.
Easy handling and maintenance: SmartFAST can pass through standard 800 mm wide door openings thanks to the overall depth of just 652 mm. All service operations are available from the front of the cabinet. Useful snap hinges keep in open position the frontal tempered glass increasing the height of the frontal opening. This solution allows the entrance of devices (such as microscopes) beneath laminar flow.
Electrical socket fitted as standard.
Specifications
Specifications
H14 Hepa
Stainless Steel