Learn More
Epredia™ SlideMate™ AS Slide Printer
Reduce specimen identification errors while enhancing lean workflow with the Epredia™ SlideMate™ AS On-Demand Slide Printer.
Brand: Epredia™ A83900002
This item is not returnable.
View return policy
Description
The SlideMate AS is the next generation Epredia Slide Printer delivering a smaller footprint than previous models, making it even easier to fit next to the microtome. The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.- Fully integrated color touch-screen and simple software are easy to use
- Build-in barcode scanner reads cassette information and imports the data to the slide label
- Connects easily to other computers or Laboratory Information Systems
- Prints directly onto the slide, eliminating the need for hand writing or labels
Specifications
SlideMate AS Basic Printing System | |
SlideMate AS basic printer with slide delivery system, color touchscreen, USB connection, power cord and simple software to print externally generated slide label from an image file. |