missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Phase Separator Papers for separation of Aqueous and Organic Phases using Grade 480

Grade 480 is impregnated with stabilized silicon, thus rendering it hydrophobic: It retains water, but allows solvents to flow through. The flow stops automatically when the entire solvent has passed through. In many applications, this phase separator paper eliminates the need to use separating funnels.

Brand:  Sartorius FT-3-602-125

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 10758421

  • £44.60 / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • This special paper separates aqueous from organic phases
  • The grade 480 is impregnated with a stabilized silicon, thus rendering it hydrophobic
  • It retains water and allows solvents to flow through
  • In many applications, this phase separator paper eliminates the need to use separating funnels
  • They are available in discs and folded filters
Specifications

Specifications

Silicone
100 Pack
4.92 in.
Separation of aqueous and organic phases
Phase Separating Paper
12.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers