Learn More
Sartorius Minisart™ NY Syringe Filters: Nonsterile
For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convenience.
£117.00 - £1420.00
Specifications
Disposable | Single-use |
---|---|
Sterility | Non-sterile |
For Use With (Application) | For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convinience |
Outlet Style | Luer Slip |
Type | Syringe Filter |
Product Code | Brand | Diameter (Metric) | Material | Pore Size | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Material | Pore Size | Quantity | Price | Quantity & Availability | ||
11786035
|
Sartorius
1776B----------K |
15 mm | Polyamide (Nylon) | 0.2 μm | 50/Pk. |
£117.00
Pack of 50 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
11710826
|
Sartorius
1776B----------Q |
15 mm | Polyamide (Nylon) | 0.2 μm | 500/Pk. |
£835.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11796035
|
Sartorius
1776C----------K |
15 mm | Polyamide (Nylon) | 0.45 μm | 50/Pk. |
£117.00
Pack of 50 |
In Stock
Log in to see stock. |
||
11720826
|
Sartorius
1776C----------Q |
15 mm | Polyamide (Nylon) | 0.45 μm | 500/Pk. |
£890.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11768154
|
Sartorius
17846----------Q |
25 mm | Polyamide (Nylon) | 0.45 μm | 500/Pk. |
£900.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11748154
|
Sartorius
17845----------Q |
25 mm | Polyamide (Nylon) | 0.2 μm | 500/Pk. |
£900.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11770876
|
Sartorius
17845----------R |
25 mm | Polyamide (Nylon) | 0.2 μm | 1000/Pk. |
£1420.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
11706045
|
Sartorius
1784B----------K |
25 mm | Polyamide (Nylon) Filter, Glass Fiber Prefilter | 0.2 μm | 500/Pk. |
£177.00
Pack of 50 |
In Stock
Log in to see stock. |
||
11780876
|
Sartorius
17846----------R |
25 mm | Polyamide (Nylon) | 0.45 μm | 1000/Pk. |
£1420.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
11790826
|
Sartorius
1784B----------Q |
25 mm | Polyamide (Nylon) Filter, Glass Fiber Prefilter | 0.2 μm | 1000/Pk. |
£1260.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11716045
|
Sartorius
1784C----------K |
25 mm | Polyamide (Nylon) Filter, Glass Fiber Prefilter | 0.45 μm | 50/Pk. |
£177.00
Pack of 50 |
In Stock
Log in to see stock. |
||
11700836
|
Sartorius
1784C----------Q |
25 mm | Polyamide (Nylon) Filter, Glass Fiber Prefilter | 0.45 μm | 500/Pk. |
£1260.00
Pack of 500 |
In Stock
Log in to see stock. |
||
Description
For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convenience.
Ready-to-use units, which offer flow rates at low inlet pressures, make rapid filtration possible. Fitted on a standard syringe.
- Particle removal
- Easy to handle
- Proven quality
- 100% tested during manufacture
Specifications
Single-use | |
For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convinience | |
Syringe Filter |
Non-sterile | |
Luer Slip |