missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S Murray™ SAMCOSyringe Luer Lock, 1 mL
SAMCOSyringes are made of glass with metal luer lock tip.
Brand: S Murray™ G813/01
19.11 GBP valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.02700kg
Description
SAMCOSyringes are made of glass with metal luer lock tip.
- Fully Interchangeable
- Italian Origin
Specifications
Metal (Tip), Neutral Glass | |
1 mL | |
Yes |