Learn More
Invitrogen™ Human ROCK1, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic.
Specifications
Specifications
| Accession Number | Q13464 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris with 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 50% glycerol, 2mM DTT and no preservative; pH 7.5 |
| Gene ID (Entrez) | 6093 |
| Molecular Weight (g/mol) | 89.7 kDa |
| Name | Human ROCK1, GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.