missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Richard-Allan Scientific™ Saccomanno Fluid

Store specimens for weeks with Epredia™ Richard-Allan Scientific™ Saccomanno Fluid, which acts as an excellent general cytological fixative.

Brand:  Epredia™ 76161

Code : A0

Additional Details : Weight : 13.30000kg Transport : UN number : 1170 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 10046548

  • £119.00 / 14 L
Estimated Shipment: 15-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Prevents air drying artifacts
  • For upper respiratory tract cytology, most notably sputum
  • Each prefilled container is labeled with a do not drink warning
  • Clear containers and tinted solution make specimen identification easy
  • Ready-to-use cytology fixative is an alcohol and polyethylene glycol (Carbowax™) solution
Specifications

Specifications

3.79L (1 Gal.)
Fixative
1 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • saccomanno fluid

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers