Learn More
Restek™ Polishing Kit for Photoionization Lamps
Used to clean the outside of the PID lamp window to restore the output.
Brand: Restek™ 20674
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Includes iron oxide cleaning compound, swabs and instruction.Specifications
PID Lamp Polishing Kit | |
Photoionization lamp | |
Iron Oxide cleaning compound, swabs and instructions |