Learn More
Restek™ Photoionization Lamps
Higher output makes it ideal for detection of BTEX compounds.
Brand: Restek™ 23020
Code : Z3
Additional Details : Weight : 1.00000kg
Description
UV energy emitting PID lamps for use with ELCD detectors.Specifications
Photoionization Lamp | |
HNU and SRI detectors | |
10/10.6 eV |