missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Mono-Lex System

Rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.

Brand:  Thermo Scientific™ R421021

Code : 50

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 12938818

  • £181.00 / 100 tests
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Thermo Scientific™ Remel Mono-Lex System is a rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.

Specifications

Specifications

2°C to 8°C
Mononucleosis Antibodies
Reagent
100 Tests/Kit
Plasma, Serum
100 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers