missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit

illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.

Brand:  Cytiva 25-6603-24

384.98 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.



Code : GH

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12987160

  • £435.00 / Each
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva.

  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).

Specifications

Specifications

Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons
Genomic DNA Amplification
24
> 10 kb
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
1 x 2.6mL 2X Denaturation Buffer, 3 x 8-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit 24 Reactions
4 h
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers