Learn More
R&D Systems™ Human PD-L2/B7-DC (NP_079515) VersaClone cDNA
Programmed Death-1 Ligand-2
Brand:  R&D Systems™ RDC1133
Additional Details : Weight : 0.00970kg
Product Code. 18431520
  • £267.81 / 10µg
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
The Human PD-L2/B7-DC (NP_079515) VersaClone cDNA clone contains the complete ORF for PD-L2/B7-DC/PDCD1LG2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PD-L2/B7-DC/PDCD1LG2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
PD-L2/B7-DC/PDCD1LG2
80380
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
NP_079515
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans