missing translation for 'onlineSavingsMsg'
Learn More

Phosphorous acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7080507
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
13598-36-2
H3PO3
MFCD00137258
Phosphonic acid
This item is not returnable. View return policy
13598-36-2
H3PO3
MFCD00137258
Phosphonic acid

Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as a precursor in the preparation of phosphine and as an intermediate in the preparation of other phosphorus compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance White crystals
Color White
Melting Point 74°C
Boiling Point 200°C (decomposition)
Density 1.651 g/cm3 at 20°C
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Assay Percent Range 97%
Quantity 500 g
Physical Form Crystals
UN Number UN2834
Merck Index 14,7346
Solubility Information Soluble in water and alcohol.
Formula Weight 82
Percent Purity 97%
Chemical Name or Material Phosphorous acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'