Learn More
Macherey-Nagel™ Phenolphthalein pH Paper
Macherey-Nagel Phenolphthalein pH Paper, Color Indicator: White to Red, pH Range: 8.3 to 10, Format: Reel, For Use With: Determination for neutralization analyses, Quantity: 1Each
Brand:  Macherey-Nagel™ 90712
Additional Details : Weight : 0.00900kg
Product Code. 11306524
  • £14.10 / 1 roll
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
Macherey-Nagel Phenolphthalein pH Paper, Color Indicator: White to Red, pH Range: 8.3 to 10, Format: Reel, For Use With: Determination for neutralization analyses, Quantity: 1Each
Specifications
Specifications
White to Red
Reel
1 Each
8.3 to 10
Determination for neutralization analyses
pH Paper