Learn More
Proteintech Phantom Dye Red 780 Viability Dye
Phantom Dye Red 780 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Brand: Proteintech PD00002-100TESTS
Additional Details : Weight : 0.00200kg
Description
- Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
- Phantom Dye Red 780 is a water soluble red fluorescent dye excited by the red laser (633 nm) making it an excellent addition to your multicolor panel.
Specifications
Flow Cytometry | |
780 Emission | |
100 Tests |