Learn More
Invitrogen™ eBioscience™ Phalloidin eFluor™ 570 Solution
Can be used to label cells of multiple species via immunohistochemical staining. eBioscience™ Phalloidin eFluor™ 570 Solution is supplied as a frozen liquid which, when thawed, forms a 1000x stock solution that can be used to prepare a 1x working solution.
Brand: Invitrogen™ 41-6559-05
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends. Because F-actin polymers compose the cytoskeleton of almost all eukaryotic cells, phalloidin conjugates can be useful probes for visualizing the structure of live or fixed cells using microscopy. Phalloidin eFluor 570 can be used to label cells of multiple species.
Phalloidin eFluor 570 is supplied as a frozen liquid. To prepare the 1000X stock solution, thaw the vial, add 30μL high-quality anhydrous DMSO, mix and briefly spin down. The resulting solution (phalloidin DMSO stock) should be aliquoted, protected from light and moisture, and stored at -20°C. To prepare a 1X working solution of phalloidin conjugate, add 1μL of 1000X phalloidin DMSO stock solution to 1mL of PBS containing 1% BSA.
Staining with Phalloidin eFluor 570 may be performed at the same time as fluorophore-conjugated antibodies. For best results, visualize phalloidin-stained cells within 1 hour of staining.
Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stabilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends. Because F-actin polymers compose the cytoskeleton of almost all eukaryotic cells, phalloidin conjugates can be useful probes for visualizing the structure of live or fixed cells using microscopy.
- Phalloidin eFluor™ 570 is supplied as a frozen liquid
- To prepare the 1000X stock solution, thaw the vial, add 30μL high-quality anhydrous DMSO, mix and briefly spin down
- The resulting solution (phalloidin DMSO stock) should be aliquoted, protected from light and moisture, and stored at −20°C
- To prepare a 1X working solution of phalloidin conjugate, add 1μL of 1000X phalloidin DMSO stock solution to 1mL of PBS containing 1% BSA
Recommended Storage
Store at ≤−20°C. Protect from light and moisture.
Specifications
Immunocytochemistry, Immunohistochemical Staining, Microscopy | |
1000X | |
≤-20°C. Protect from light and moisture. | |
Frozen Liquid |
300 Tests | |
A peptide toxin isolated from the Amanita phalloides mushroom | |
Phalloidin eFluor 570 Solution |