missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4.01 Calibration Buffer Solution Sachets, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Packaging:
25Pack
Description
- Premium quality pH 4.01 calibration buffer that is NIST traceable and supplied with a Certificate of Analysis
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- pH 4.01 calibration buffer produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionized water, certified weight-checked balances, and Class A glassware
- In a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
- Ideal for users that do not calibrate frequently
- Ideal for field use due to portability
Specifications
Color | Undesignated |
pH | 4.01 |
Physical Form | Liquid |
Quantity | 20 mL |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
Packaging | 25Pack |
Traceability to NIST | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'