Learn More
Petroleum Ether a.r. For Laboratory Use, Chem Lab
Brand: Chem Lab CL00.1608.5011
Additional Details : CAS Number : 64742-49-0 Weight : 5.00000kg Transport : UN number : 1268 Chem class : 3 Pack group : II
Specifications
-100°C | |
Colorless | |
40°C to 60°C | |
-21 °C | |
Liquid | |
Analytical Reagent |
64742-49-0 | |
0.653 g/mL | |
5 L | |
Fluorinated HDPE | |
< 0.01% |
Safety and Handling
- Petroleum ether 40-60
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Petroleum ether