Learn More
Thermo Scientific™ Peristaltic Pump Tubing for iCAP™ Q/Qnova Series ICP-MS Systems
Designed for durability and fitness for purpose, a range of Thermo Scientific™ Peristaltic Pump Tubing is available to suit your sample matrices and ICP-MS applications.
Brand: Thermo Scientific™ 1320110
Additional Details : Weight : 0.03500kg
Description
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.
Select High Quality Consumables for Improved PerformanceChoose from the following Peristaltic Pump Tubing:
- Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
- Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
- Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
- Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
- Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
- Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene
Specifications
Peristaltic Pump Tubing | |
Santoprene, Dia. 1.295, Gray/Gray | |
iCAP Q/Qnova Series ICP-MS Systems |