missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluoro-2-methyl-2-pentene, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Specifications
CAS | 1584-03-8 |
Color | Colorless to Yellow |
Density | 1.6200g/mL |
Boiling Point | 53.0°C to 61.0°C |
Flash Point | −12°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C6F12 |
Linear Formula | C2F5CF=C(CF3)2 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'