missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
67.54 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
ClHO4, CAS Number-7601-90-3, unii-v561v90bg2, ueberchlorsaeure, perchlorsaeure, perchloric acid, hclo4, hyperchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchloric acid, 1L, CHEBI:29221, Colorless, VLTRZXGMWDSKGL-UHFFFAOYSA-N, 11325, 100.454
Specifications
pH | 0.1 |
Color | Colorless |
Packaging | HDPE plastic bottle |
Quantity | 1 L |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'