missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex IFN-beta Human ELISA Kit
Click to view available options
Quantity:
96 Assays
Unit Size:
96 tests
Description
Includes
Pre-coated plate, standard, controls, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Detemines levels of human IFN-β in serum, plasma, supernatant, and urine.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
Specifications
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Application) | Colorimetric |
| Gene ID (Entrez) | 3456 |
| Kit Contents | Chromogen, Controls, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
| Precision | I IU/mL |
| Quantity | 96 Assays |
| Regulatory Status | For research use only (RUO) |
| Target | Interferon, beta 1, fibroblast |
| Test Time | 2.5 hr. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction