missing translation for 'onlineSavingsMsg'
Learn More

Nicotine Bitartrate Dihydrate, MP Biomedicals™

Product Code. p-4405003 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg

Product Code. 11482902

Brand: MP Biomedicals 0215355483

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

Molecular Formula C18H30N2O14
Molecular Weight (g/mol) 498.44
MDL Number MFCD00150899
InChI Key LDMPZNTVIGIREC-HYRPJEHINA-N
SMILES O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1

Specifications

CAS 6-3-6019
Melting Point 93°C to 95°C (literature)
Color White
pH 3.18 (10% aq. soln.) (literature)
Quantity 250 mg
Synonym nicotine ditartrate dihydrate, unii-r7m676m8yv, nicotine bitartrate usan, 2r,3r-2,3-dihydroxybutanedioic acid;3-2s-1-methylpyrrolidin-2-yl pyridine;dihydrate, nicotine bitartrate who-dd, nicotine acid tartrate dihydrate, s-3-1-methylpyrrolidin-2-yl pyridine bis 2r,3r-2,3-dihydroxysuccinate dihydrate, 2r,3r-2,3-dihydroxybutanedioic acid; 3-2s-1-methylpyrrolidin-2-yl pyridine; dihydrate, 3-2s-1-methyl-2-pyrrolidinyl pyridine, 2r,3r-2,3-dihydroxybutanedioate 1:2 , dihydrate
Solubility Information Soluble in water (50mg/mL - clear to slightly hazy, colorless to faint yellow solution) or alcohol.
IUPAC Name bis((2R,3R)-2,3-dihydroxybutanedioic acid) 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine dihydrate
Formula Weight 498.44
Physical Form Powder
Chemical Name or Material Nicotine Bitartrate Dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.